Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cytl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081106.1 | NP_001074575.1 | 4 | 409 | ||
GenBank mRNA | AK076770.1 | - | 3 | 301 | ||
BC063103.1 | - | 4 | 369 | |||
BC116224.1 | - | 4 | 383 | 40091368 | ||
BC116225.1 | - | 4 | 383 | 40091369 | ||
EF108311.1 | - | 4 | 380 |