Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GPRC5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003979.3 | NP_003970.1 | 4 | 2636 | ||
GenBank mRNA | AF095448.1 | - | 4 | 2085 | ||
AF506289.1 | - | 4 | 2093 | |||
AK001761.1 | - | 4 | 2239 | |||
AK122672.1 | - | 4 | 2876 | |||
AK172760.1 | - | 3 | 1606 | |||
BC003665.2 | - | 4 | 2081 | 2988011 |
Set Membership: |
![]() |