Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tmf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081111.2 | NP_001074580.1 | 17 | 4465 | ||
GenBank mRNA | AK078561.1 | - | 1 | 801 | ||
AK136794.1 | - | 1 | 808 | |||
AK161423.1 | - | 1 | 799 |