Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Prr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175022.2 | NP_778187.2 | 4 | 2941 | ||
GenBank mRNA | AK173130.1 | - | 1 | 2380 | ||
BC058674.1 | - | 1 | 1580 | |||
BC075628.1 | - | 5 | 1850 |