Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Setdb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081024.1 | NP_001074493.1 | 15 | 2945 | ||
GenBank mRNA | EU155090.1 | - | 14 | 2759 | ||
EU155091.1 | - | 14 | 2759 | |||
EU155092.1 | - | 14 | 2777 | |||
EU155093.1 | - | 13 | 2693 | |||
EU155094.1 | - | 13 | 2693 | |||
EU155096.1 | - | 13 | 2416 | |||
EU155097.1 | - | 15 | 2818 | |||
EU155099.1 | - | 14 | 2775 |