Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gp6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163014.1 | NP_001156486.1 | 6, 7 | 770 | ||
GenBank mRNA | BC138170.1 | - | 6, 7 | 754 | 8861189 |