Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psma4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017281.1 | NP_058977.1 | 8 | 648 | ||
GenBank mRNA | BC063170.1 | - | 8 | 668 | 6921964 | |
FQ212536.1 | - | 4 | 281 | |||
FQ213330.1 | - | 4 | 281 | |||
FQ215861.1 | - | 4 | 281 | |||
FQ216011.1 | - | 4 | 281 | |||
FQ216241.1 | - | 4 | 281 | |||
FQ220234.1 | - | 4 | 280 | |||
FQ220280.1 | - | 4 | 281 | |||
FQ226777.1 | - | 4 | 281 | |||
FQ234380.1 | - | 4 | 281 | |||
X53304.1 | - | 8 | 648 | |||
X55986.1 | - | 8 | 604 |