Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Serpine3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199945.1 | NP_001186874.1 | 3 | 419 | ||
GenBank mRNA | AK053602.1 | - | 3 | 421 |