Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Acox1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017340.2 | NP_059036.1 | 245 | |||
GenBank mRNA | BC085743.1 | - | 245 | 7109722 | ||
J02752.1 | - | 228 |
Gene Symbol: LOC100910385 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750965.2 | XP_003751013.1 | 2 | 239 |