Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ttll10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029264.2 | NP_083540.1 | 7 | 766 | ||
GenBank mRNA | AK014686.1 | - | 5 | 1063 | ||
AK016396.1 | - | 4 | 355 | |||
AK133079.1 | - | 7 | 767 | |||
AM690754.1 | - | 4 | 650 |