Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TPM1 SIRNA Assays ›
Gene Symbol:
Tpm1
Gene Name:
tropomyosin 1, alpha
Gene Aliases:
Alpha-tm, Tma2, Tmsa
Chromosome Location:
Chr.8: 72814737 - 72841496 on Build Rnor_6.0
UniGene ID:
Rn.87540
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
50392
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tpm1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001034068.1
NP_001029240.1
3, 4
490
NM_001034069.1
NP_001029241.1
3, 4
490
NM_001034070.1
NP_001029242.1
3, 4
490
NM_001034071.1
NP_001029243.1
3, 4
490
NM_001034072.1
NP_001029244.1
3, 4
490
NM_001034073.1
NP_001029245.1
2, 3
427
NM_001034074.1
NP_001029246.1
2, 3
427
NM_001034075.1
NP_001029247.1
2, 3
427
NM_001301336.1
NP_001288265.1
3, 4
490
NM_001301342.1
NP_001288271.1
3, 4
490
NM_001301736.1
NP_001288665.1
2, 3
427
NM_019131.2
NP_062004.1
2, 3
427
XM_006243386.2
XP_006243448.1
3, 4
482
XM_006243388.2
XP_006243450.1
3, 4
482
XM_006243389.2
XP_006243451.1
3, 4
482
XM_006243390.2
XP_006243452.1
3, 4
482
XM_006243393.1
XP_006243455.1
2, 3
425
XM_006243394.1
2, 3
443
XM_006243395.2
XP_006243457.1
2, 3
434
XM_008766207.1
XP_008764429.1
3, 4
482
XM_008766208.1
XP_008764430.1
2, 3
425
GenBank mRNA
AF372215.1
-
2, 3
276
AF372216.1
-
3, 4
389
AF372217.1
-
3, 4
395
BC078780.1
-
2, 3
394
7106337
BU670983.1
-
3, 4
490
CB585296.1
-
2, 3
430
CF108680.1
-
3, 4
438
CK483405.1
-
3, 4
444
CO562261.1
-
457
FQ074026.1
-
3, 4
548
FQ214619.1
-
3, 4
465
FQ214803.1
-
3, 4
461
FQ214809.1
-
3, 4
459
FQ214835.1
-
3, 4
459
FQ214864.1
-
3, 4
463
FQ214868.1
-
3, 4
459
FQ214869.1
-
3, 4
465
FQ214872.1
-
3, 4
458
FQ214877.1
-
3, 4
459
FQ214894.1
-
3, 4
465
FQ214902.1
-
3, 4
459
FQ214911.1
-
3, 4
461
FQ214944.1
-
3, 4
465
FQ214946.1
-
3, 4
465
FQ214991.1
-
3, 4
459
FQ215186.1
-
3, 4
465
FQ215188.1
-
3, 4
459
FQ215206.1
-
3, 4
494
FQ215226.1
-
3, 4
465
FQ215459.1
-
3, 4
461
FQ215512.1
-
3, 4
465
FQ215522.1
-
3, 4
536
FQ215524.1
-
3, 4
461
FQ215538.1
-
3, 4
465
FQ215551.1
-
3, 4
459
FQ215698.1
-
3, 4
459
FQ215703.1
-
3, 4
459
FQ215724.1
-
3, 4
546
FQ215746.1
-
3, 4
461
FQ215761.1
-
3, 4
459
FQ215768.1
-
3, 4
459
FQ215803.1
-
3, 4
465
FQ215808.1
-
3, 4
461
FQ215815.1
-
3, 4
459
FQ215856.1
-
3, 4
459
FQ215919.1
-
3, 4
467
FQ215944.1
-
3, 4
466
FQ215969.1
-
3, 4
459
FQ215995.1
-
3, 4
461
FQ216014.1
-
3, 4
543
FQ216018.1
-
3, 4
465
FQ216052.1
-
3, 4
459
FQ216105.1
-
3, 4
459
FQ216118.1
-
3, 4
543
FQ216145.1
-
3, 4
465
FQ216160.1
-
3, 4
458
FQ216319.1
-
3, 4
461
FQ216340.1
-
3, 4
465
FQ216368.1
-
3, 4
461
FQ216400.1
-
3, 4
461
FQ216494.1
-
3, 4
459
FQ216552.1
-
3, 4
465
FQ216557.1
-
3, 4
465
FQ216569.1
-
553
FQ216615.1
-
3, 4
374
FQ216637.1
-
3, 4
461
FQ216683.1
-
3, 4
461
FQ216697.1
-
3, 4
461
FQ216747.1
-
3, 4
461
FQ217468.1
-
2, 3
210
FQ217503.1
-
3, 4
465
FQ217616.1
-
1, 2
27
FQ217637.1
-
3, 4
465
FQ217808.1
-
3, 4
461
FQ223706.1
-
3, 4
461
FQ223993.1
-
3, 4
463
FQ224036.1
-
3, 4
376
FQ224207.1
-
3, 4
459
FQ224838.1
-
3, 4
465
FQ224943.1
-
3, 4
463
FQ224961.1
-
3, 4
461
M34134.1
-
2, 3
412
M34135.1
-
3, 4
424
M34136.1
-
1, 2
142
M60666.1
-
3, 4
437
M60667.1
-
3, 4
437
M60668.1
-
2, 3
374
M60669.1
-
2, 3
374
X02411.1
-
3, 4
388
X02412.1
-
1, 2
144
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_001164255, NM_001164254, NM_001164256, NM_001164250, NM_024427, and others.
Panther Classification:
Molecular Function -
actin binding motor protein
Gene Ontology Categories:
Function(s)
Process(es)
in utero embryonic development
positive regulation of heart rate by epinephrine
muscle contraction
muscle filament sliding
negative regulation of cell migration
ruffle organization
positive regulation of ATPase activity
cellular response to reactive oxygen species
wound healing
regulation of ATPase activity
sarcomere organization
positive regulation of cell adhesion
positive regulation of stress fiber assembly
actin filament capping
ventricular cardiac muscle tissue morphogenesis
cardiac muscle contraction
actin binding
cytoskeletal protein binding
protein homodimerization activity
protein N-terminus binding
actin filament binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0