Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033432.3 | NP_001028604.1 | 4 | 3340 | ||
GenBank mRNA | AK132340.1 | - | 4 | 3341 | ||
BC094026.1 | - | 3 | 1954 |