Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chrng | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019145.1 | NP_062018.1 | 4 | 426 | ||
GenBank mRNA | X74834.1 | - | 4 | 426 |