Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mroh7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126487.1 | NP_001119959.1 | 19 | 3380 | ||
GenBank mRNA | AK134749.1 | - | 10 | 1802 |