Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: 6430531B16Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033465.2 | NP_001028637.1 | 3 | 412 | ||
GenBank mRNA | AK131789.1 | - | 3 | 413 | ||
AK164921.1 | - | 3 | 382 | |||
BC025923.1 | - | 2 | 748 | |||
BC145460.1 | - | 3 | 376 | |||
BC145461.1 | - | 3 | 376 | |||
BC145730.1 | - | 3 | 376 | 40131033 | ||
BC171930.1 | - | 3 | 376 |