Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hapln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019189.2 | NP_062062.2 | 3 | 225 | ||
XM_006231748.2 | XP_006231810.1 | 3 | 426 | |||
XM_008760634.1 | XP_008758856.1 | 2 | 635 | |||
GenBank mRNA | BC128740.1 | - | 3 | 225 | 8371477 |