Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ndufa4l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098789.1 | NP_001092259.1 | 4 | 387 | ||
GenBank mRNA | AK131609.1 | - | 4 | 359 | ||
AK131662.1 | - | 4 | 356 | |||
BC064011.1 | - | 4 | 348 | |||
BC099496.1 | - | 4 | 348 | 30847512 |