Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Mpdz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019196.1 | NP_062069.1 | 4, 5 | 587 | ||
XM_006238340.2 | XP_006238402.1 | 4, 5 | 627 | |||
XM_006238341.2 | XP_006238403.1 | 4, 5 | 627 | |||
XM_006238342.2 | XP_006238404.1 | 4, 5 | 627 | |||
XM_006238343.2 | XP_006238405.1 | 4, 5 | 627 | |||
XM_006238344.2 | XP_006238406.1 | 4, 5 | 627 | |||
XM_008763763.1 | XP_008761985.1 | 4, 5 | 1796 | |||
XM_008763764.1 | XP_008761986.1 | 4, 5 | 628 | |||
GenBank mRNA | AJ001320.1 | - | 4, 5 | 587 |