Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sfta2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163194.1 | NP_001156666.1 | 2 | 244 | ||
GenBank mRNA | AK087178.1 | - | 2 | 245 |