Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apcdd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133237.3 | NP_573500.2 | 3 | 656 | ||
GenBank mRNA | AK156319.1 | - | 3 | 657 | ||
AK172004.1 | - | 660 | ||||
AK190159.1 | - | 2 | 130 | |||
AK194715.1 | - | 2 | 130 | |||
BC141269.1 | - | 4 | 430 | 9055881 | ||
BC141270.1 | - | 4 | 476 | 9055882 |
Gene Symbol: Gars | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK190416.1 | - | 365 |