Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Wdr70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081402.1 | NP_001074871.1 | 14 | 1538 | ||
GenBank mRNA | AK159714.1 | - | 14 | 1520 | ||
AK160933.1 | - | 14 | 1475 |