- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Zfp260 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017364.4 | NP_059060.1 | 2 | 1100 | ||
XM_008759183.1 | XP_008757405.1 | 1467 | ||||
GenBank mRNA | BC099805.1 | - | 1 | 947 | 7937786 | |
U56862.1 | - | 1 | 938 |