Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm7120 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039244.3 | NP_001034333.2 | 2 | 207 | ||
NM_001177666.1 | NP_001171137.1 | 2 | 180 | |||
GenBank mRNA | AK050607.1 | - | 2 | 208 | ||
AK162713.1 | - | 208 | ||||
BC021422.1 | - | 2 | 189 | 5011629 | ||
BC103784.1 | - | 1 | 183 | 6309338 |