Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sdr39u1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001082975.1 | NP_001076444.1 | 4 | 245 | ||
GenBank mRNA | AK009339.1 | - | 4 | 225 | ||
AK049709.1 | - | 4 | 239 | |||
BC058857.1 | - | 2 | 418 | |||
BC087941.1 | - | 4 | 224 | 30303350 |