Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Polr2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025554.2 | NP_079830.2 | 7 | 964 | ||
GenBank mRNA | AK010567.1 | - | 7 | 972 | ||
AK015823.1 | - | 7 | 962 | |||
AK088610.1 | - | 7 | 1021 | |||
AK132109.1 | - | 7 | 1019 | |||
AK144668.1 | - | 7 | 1025 | |||
AK145201.1 | - | 7 | 1030 | |||
BC005599.1 | - | 7 | 1016 | |||
BC026842.1 | - | 5 | 1192 | 4210827 | ||
BC037681.1 | - | 4 | 2359 | |||
BC045521.1 | - | 7 | 964 | 2647754 |