Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Edem3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039644.2 | NP_001034733.2 | 20 | 3430 | ||
GenBank mRNA | BC022925.1 | - | 1 | 290 | ||
BC060718.1 | - | 14 | 2532 |