Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vma21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081356.3 | NP_001074825.1 | 3 | 529 | ||
NM_001290780.1 | NP_001277709.1 | 3 | 656 | |||
NM_001290781.1 | NP_001277710.1 | 4 | 830 | |||
GenBank mRNA | AJ748652.1 | - | 3 | 477 | ||
AK008848.1 | - | 3 | 487 | |||
AK011632.1 | - | 3 | 495 | |||
AK045401.1 | - | 3 | 487 | |||
AK049296.1 | - | 3 | 467 | |||
AK049488.1 | - | 3 | 500 | |||
AK164163.1 | - | 3 | 484 | |||
BC028317.1 | - | 3 | 467 | |||
BC094639.1 | - | 3 | 483 | 1361961 |