Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Parp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163575.1 | NP_001157047.1 | 4 | 669 | ||
NM_001163576.1 | NP_001157048.1 | 3 | 773 | |||
GenBank mRNA | AK028373.1 | - | 4 | 617 | ||
AK035309.1 | - | 4 | 663 | |||
AK046755.1 | - | 3 | 754 | |||
AK146336.1 | - | 4 | 670 | |||
AK153205.1 | - | 3 | 510 | |||
AK166291.1 | - | 4 | 620 | |||
BC024074.1 | - | 4 | 626 |