Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026032.1 | NP_080308.1 | 4 | 768 | ||
GenBank mRNA | AK006006.1 | - | 2 | 503 | ||
AK012814.1 | - | 2 | 503 | |||
AK019398.1 | - | 2 | 498 | |||
AK041290.1 | - | 4 | 768 | |||
BC021460.1 | - | 4 | 1051 | 5054997 |