Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cwc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026072.1 | NP_080348.1 | 11 | 1218 | ||
GenBank mRNA | AK014025.1 | - | 11 | 1210 | ||
AK075902.1 | - | 11 | 1196 | |||
AK077779.1 | - | 11 | 1161 | |||
AK166770.1 | - | 11 | 1218 | |||
BC025437.1 | - | 11 | 1018 | 5025084 |