Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Camsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081360.1 | NP_001074829.1 | 19 | 5135 | ||
GenBank mRNA | AK081975.1 | - | 5 | 751 | ||
AK122436.1 | - | 7 | 2475 | |||
BC138367.1 | - | 18 | 4711 | 8861387 | ||
BC145258.1 | - | 17 | 4678 | 9053811 |