Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdzd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040136.2 | NP_001035226.1 | 4 | 1324 | ||
NM_001040137.2 | NP_001035227.1 | 4 | 1094 | |||
GenBank mRNA | AK005921.1 | - | 4 | 1325 | ||
AK006727.1 | - | 4 | 1228 | |||
AK015115.1 | - | 4 | 1095 | |||
AK017019.1 | - | 4 | 1232 | |||
BC147551.1 | - | 4 | 1320 | 9056542 | ||
BC147563.1 | - | 4 | 1320 | 9056553 | ||
BC172089.1 | - | 3 | 1130 |