Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rere | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085492.1 | NP_001078961.1 | 23 | 7299 | ||
GenBank mRNA | AK004046.1 | - | 1 | 771 | ||
AK122287.1 | - | 6 | 4982 | |||
AK169818.1 | - | 1 | 1190 | |||
BC110693.1 | - | 4 | 2911 |