Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Flnc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081185.1 | NP_001074654.1 | 33 | 5776 | ||
GenBank mRNA | BC052186.1 | - | 6 | 725 | ||
BC060276.1 | - | 12 | 1837 | |||
BC151097.1 | - | 33 | 5581 | 9088010 | ||
BC158128.1 | - | 32 | 5482 | 9007396 | ||
BC172159.1 | - | 32 | 5482 |