Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027009.2 | NP_081285.1 | 5, 6 | 631 | ||
GenBank mRNA | AK042393.1 | - | 5, 6 | 632 | ||
AK145126.1 | - | 5, 6 | 658 | |||
AK165891.1 | - | 5, 6 | 626 | |||
AK167047.1 | - | 5, 6 | 632 | |||
AK209135.1 | - | 2, 3 | 208 | |||
BC026795.1 | - | 5, 6 | 602 | 4015134 |