Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhobtb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081347.1 | NP_001074816.1 | 6 | 1413 | ||
NM_001252636.1 | NP_001239565.1 | 6 | 1396 | |||
NM_001252637.1 | NP_001239566.1 | 6 | 1517 | |||
NM_001252638.1 | NP_001239567.1 | 7 | 1576 | |||
GenBank mRNA | AK005770.1 | - | 6 | 1396 | ||
AK034826.1 | - | 6 | 1413 | |||
AK078306.1 | - | 5 | 1350 | |||
AK173015.1 | - | 1 | 777 | |||
BC099973.1 | - | 6 | 1394 | 6432524 | ||
BC137646.1 | - | 5 | 1347 | 8860665 | ||
BC137647.1 | - | 5 | 1347 | 8860666 | ||
BC144800.1 | - | 6 | 1406 | 9053348 |