Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nt5dc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027289.1 | NP_081565.1 | 7 | 508 | ||
GenBank mRNA | BC011230.1 | - | 7 | 508 | 4207917 |