Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dgkb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019304.1 | NP_062177.1 | 3 | 383 | ||
XM_008764649.1 | XP_008762871.1 | 2 | 324 | |||
XM_008764650.1 | XP_008762872.1 | 2 | 324 | |||
XM_008764651.1 | XP_008762873.1 | 2 | 322 | |||
GenBank mRNA | D16100.1 | - | 3 | 383 |