Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081368.1 | NP_001074837.1 | 5 | 1286 | ||
GenBank mRNA | AK039905.1 | - | 5 | 1111 | ||
AK040787.1 | - | 6 | 1339 | |||
AK133804.1 | - | 5 | 1227 | |||
AK184985.1 | - | 1 | 97 | |||
AK207313.1 | - | 1 | 97 | |||
BC029679.1 | - | 670 | ||||
BC082558.1 | - | 5 | 1255 | 30605020 |