Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbccd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081368.1 | NP_001074837.1 | 6 | 1336 | ||
GenBank mRNA | AK039905.1 | - | 6 | 1161 | ||
AK040787.1 | - | 7 | 1389 | |||
AK133804.1 | - | 6 | 1277 | |||
AK207313.1 | - | 2 | 147 | |||
BC082558.1 | - | 6 | 1305 | 30605020 |