Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Naa40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027643.1 | NP_081919.1 | 8 | 2678 | ||
GenBank mRNA | AK033510.1 | - | 8 | 2500 | ||
AK033896.1 | - | 8 | 2509 | |||
AK035418.1 | - | 7 | 3396 | |||
AK162625.1 | - | 7 | 2772 | |||
AK170548.1 | - | 8 | 2504 | |||
AK181058.1 | - | 1 | 241 | |||
AK181303.1 | - | 1 | 242 | |||
AK188291.1 | - | 1 | 241 | |||
AK217157.1 | - | 1 | 241 | |||
BB112808.2 | - | 1 | 74 | |||
BC020020.1 | - | 8 | 2529 | 3156924 | ||
BC049808.1 | - | 2 | 1916 | |||
BC058212.1 | - | 3 | 2337 |