Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdss2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168289.1 | NP_001161761.1 | 6 | 1211 | ||
NM_027772.2 | NP_082048.2 | 6 | 1211 | |||
GenBank mRNA | AB210840.1 | - | 6 | 935 | ||
AK140091.1 | - | 6 | 1098 | |||
AK165732.1 | - | 6 | 1210 | |||
BC147693.1 | - | 6 | 988 | 9056666 | ||
BC147694.1 | - | 6 | 988 | 9056667 |