Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam217b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081289.1 | NP_001074758.1 | 4 | 1157 | ||
GenBank mRNA | AK155442.1 | - | 4 | 1159 |