- Order Status
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: Cdc40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027879.2 | NP_082155.1 | 15 | 3171 | ||
GenBank mRNA | AK004569.1 | - | 15 | 3138 | ||
BC061220.1 | - | 6 | 724 |