Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc43a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081349.2 | NP_001074818.2 | 15 | 1938 | ||
NM_001083809.1 | NP_001077278.1 | 14 | 1877 | |||
NM_024497.1 | NP_078773.1 | 15 | 1836 | |||
GenBank mRNA | AB103034.1 | - | 15 | 1907 | ||
AK011417.1 | - | 15 | 1936 | |||
AK031215.1 | - | 14 | 1880 | |||
AK046758.1 | - | 15 | 1930 | |||
BC053747.1 | - | 15 | 1735 | 6330614 |