Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019295.1 | NP_062168.1 | 5 | 605 | ||
GenBank mRNA | BC098918.1 | - | 5 | 612 | 7384975 | |
D10728.1 | - | 5 | 605 | |||
X78985.1 | - | 5 | 552 |
Gene Symbol: LOC100911215 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006231081.2 | XP_006231143.2 | 426 |