Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pradc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163427.1 | NP_001156899.1 | 5 | 712 | ||
NM_028505.2 | NP_082781.1 | 4 | 544 | |||
GenBank mRNA | AK006658.1 | - | 5 | 678 | ||
BC115452.2 | - | 4 | 490 | 40100110 | ||
CJ208123.1 | - | 1 | 33 |