Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kif2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_028547.2 | NP_082823.1 | 1 | 452 | ||
GenBank mRNA | AK030188.1 | - | 1 | 453 | ||
BC100484.1 | - | 1 | 428 | 30919380 | ||
BC132056.1 | - | 1 | 506 | 40130333 | ||
BC137981.1 | - | 1 | 506 | 8861000 |