Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mipol1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164370.1 | NP_001157842.1 | 7 | 843 | ||
GenBank mRNA | AK006974.1 | - | 752 | |||
AK044766.1 | - | 6 | 570 | |||
AK077915.1 | - | 7 | 823 | |||
AK138175.1 | - | 336 | ||||
AK168989.1 | - | 6 | 628 |