Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081164.1 | NP_001074633.1 | 21 | 6033 | ||
NM_001256033.1 | NP_001242962.1 | 21 | 6036 | |||
GenBank mRNA | AK078157.1 | - | 1 | 2229 | ||
AK084296.1 | - | 1 | 1790 | |||
AK122429.1 | - | 16 | 5327 | |||
AK169183.1 | - | 1 | 830 | |||
BC042641.1 | - | 1 | 2774 | |||
BC138373.1 | - | 21 | 6008 | 8861393 | ||
BC145259.1 | - | 21 | 6005 | 9053812 |